Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145122.2 | NP_660104.2 | 4 - 5 | 683 | 75 | |
NR_028114.1 | 4 - 5 | 392 | 75 | |||
GenBank mRNA | AK020743.1 | - | 4 - 5 | 393 | 75 | |
AK143073.1 | - | 4 - 5 | 685 | 75 | ||
AK162716.1 | - | 4 - 5 | 383 | 75 | ||
BC010822.1 | - | 4 - 5 | 424 | 4218547 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |