Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21428087 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
BC010822.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
BE690943.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
NM_145122.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
NR_028114.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
AK020743.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
BC010822.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
BE690943.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
NM_145122.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
NR_028114.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P395164 |
AK020743.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
548570 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467354 |
BC008517.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467354 |
NM_009459.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467354 |
U19854.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467354 |
XM_001472265.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
467354 |
XM_001477372.1 |
refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
607424 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
151572 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135745 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
BC010822.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
BE690943.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
NM_145122.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
NR_028114.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
AK020743.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
BC010822.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
BE690943.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
NM_145122.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
NR_028114.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P395164 |
AK020743.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428130 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428114 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428133 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428117 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428118 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428128 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428092 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428106 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428096 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428125 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428115 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428121 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428113 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428087 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428119 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428127 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428123 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428111 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428122 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428126 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428091 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428098 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428094 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428129 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428109 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428086 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428090 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428105 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21428132 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135745 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P241732 |
BC010822.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P241732 |
NM_145122.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P241732 |
NR_028114.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P241732 |
AK020743.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P241732 |
AK143073.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
BC010822.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
BE690943.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
NM_145122.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
NR_028114.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
AK020743.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
BC010822.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
BE690943.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
NM_145122.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
NR_028114.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P395164 |
AK020743.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P112701 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135745 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01775440 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21428087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428066 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428128 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428222 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428106 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428048 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428013 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428010 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428042 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428034 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428132 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428108 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428219 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428006 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428030 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428129 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428062 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428125 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428014 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428076 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428147 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428123 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428130 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428150 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428067 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428039 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428122 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428068 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428096 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428208 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428054 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428051 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428133 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428015 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428055 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428070 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428213 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428078 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428046 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428074 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428018 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428075 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428205 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428114 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21428105 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428096 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428094 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428071 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428074 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428128 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428076 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428122 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428123 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428106 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428084 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727442 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428115 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727407 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428105 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428133 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428100 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727435 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727413 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727443 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727383 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428082 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428075 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727419 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727405 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727408 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428121 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428117 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428092 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428090 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428087 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428098 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428086 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428091 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428081 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428103 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727393 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21428073 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727381 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01727386 |
- |
- |