Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspear | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287074.1 | NP_001274003.1 | 4 - 5 | 879 | 64 | |
GenBank mRNA | AJ487520.1 | - | 3 - 4 | 550 | 64 | |
AK082868.1 | - | 2 - 3 | 351 | 64 | ||
JQ815565.1 | - | 4 - 5 | 879 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |