Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tubgcp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146190.2 | NP_666302.2 | 14 - 15 | 1998 | 82 | |
GenBank mRNA | AK032031.1 | - | 14 - 15 | 1958 | 82 | |
AK044887.1 | - | 14 - 15 | 1982 | 82 | ||
AK051326.1 | - | 14 - 15 | 1993 | 82 | ||
AK134963.1 | - | 6 - 7 | 1116 | 82 | ||
AK160939.1 | - | 14 - 15 | 1912 | 82 | ||
AK169443.1 | - | 5 - 6 | 1342 | 82 | ||
AK173291.1 | - | 11 - 12 | 1375 | 82 | ||
BC027075.1 | - | 6 - 7 | 1061 | 5355335 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |