Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148930.3 | NP_683732.1 | 5 - 6 | 582 | 65 | |
GenBank mRNA | AJ309168.1 | - | 4 - 5 | 417 | 65 | |
AK133999.1 | - | 5 - 6 | 540 | 65 | ||
AK134183.1 | - | 5 - 6 | 577 | 65 | ||
AK146880.1 | - | 5 - 6 | 577 | 65 | ||
BC005719.1 | - | 1 - 2 | 248 | 65 | ||
BC019766.1 | - | 5 - 6 | 527 | 65 | ||
BC023854.1 | - | 5 - 6 | 567 | 5346780 | 65 | |
BC031899.1 | - | 5 - 6 | 567 | 5362121 | 65 | |
BC043058.1 | - | 5 - 6 | 519 | 6419695 | 65 | |
BC054729.1 | - | 5 - 6 | 570 | 6419758 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |