Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148945.2 | NP_683747.1 | 5 - 6 | 739 | 130 | |
GenBank mRNA | AK079102.1 | - | 832 | 130 | ||
AK154778.1 | - | 5 - 6 | 652 | 130 | ||
AY083469.1 | - | 5 - 6 | 401 | 130 | ||
BC150156.1 | - | 5 - 6 | 404 | 130 | ||
BC150478.1 | - | 5 - 6 | 404 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |