Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Osbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152818.2 | NP_690031.2 | 9 - 10 | 2022 | 80 | |
GenBank mRNA | AK007088.1 | - | 5 - 6 | 603 | 80 | |
AK044598.1 | - | 6 - 7 | 811 | 80 | ||
BC031794.1 | - | 7 - 8 | 985 | 4501119 | 80 | |
BC051185.1 | - | 7 - 8 | 982 | 80 | ||
BC058356.1 | - | 8 - 9 | 1165 | 5696357 | 80 | |
BC058602.1 | - | 7 - 8 | 985 | 6467653 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |