Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007842.2 | NP_031868.2 | 9 - 10 | 1057 | 124 | |
GenBank mRNA | AK135603.1 | - | 4 - 5 | 371 | 124 | |
AK141824.1 | - | 9 - 10 | 1052 | 124 | ||
AK148423.1 | - | 9 - 10 | 1053 | 124 | ||
AY512925.1 | - | 5 - 6 | 513 | 124 | ||
BC089159.1 | - | 9 - 10 | 1051 | 124 | ||
U91922.1 | - | 9 - 10 | 1014 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |