Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008576.3 | NP_032602.1 | 27 - 28 | 3995 | 93 | |
GenBank mRNA | AF022908.1 | - | 27 - 28 | 3957 | 93 | |
AK029876.1 | - | 27 - 28 | 3996 | 93 | ||
AK049174.1 | - | 11 - 12 | 1762 | 93 | ||
AK140421.1 | - | 4 - 5 | 1538 | 93 | ||
BC090617.1 | - | 27 - 28 | 3966 | 30943198 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |