Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008766.3 | NP_032792.2 | 5 - 6 | 1203 | 90 | |
GenBank mRNA | AK035971.1 | - | 5 - 6 | 1021 | 90 | |
BC021647.1 | - | 5 - 6 | 1171 | 4163278 | 90 | |
U52842.1 | - | 5 - 6 | 1084 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |