Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St8sia3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009182.3 | NP_033208.2 | 4 - 5 | 1098 | 100 | |
GenBank mRNA | AK015874.1 | - | 4 - 5 | 1082 | 100 | |
AK133780.1 | - | 2 - 3 | 383 | 100 | ||
AK134678.1 | - | 2 - 3 | 394 | 100 | ||
AK161482.1 | - | 4 - 5 | 1099 | 100 | ||
BC075645.1 | - | 2 - 3 | 393 | 30620472 | 100 | |
X80502.1 | - | 2 - 3 | 383 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |