Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009531.2 | NP_033557.2 | 10 - 11 | 2102 | 61 | |
GenBank mRNA | AB071144.1 | - | 10 - 11 | 2039 | 61 | |
AK004713.1 | - | 10 - 11 | 2042 | 61 | ||
AK028595.1 | - | 10 - 11 | 2040 | 61 | ||
AK166981.1 | - | 10 - 11 | 2037 | 61 | ||
BC052725.1 | - | 10 - 11 | 2064 | 6812198 | 61 | |
U27398.1 | - | 10 - 11 | 2013 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |