Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Avil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009635.3 | NP_033765.2 | 15 - 16 | 1964 | 105 | |
GenBank mRNA | AF041448.1 | - | 15 - 16 | 1915 | 105 | |
AF059486.1 | - | 15 - 16 | 1934 | 105 | ||
AK154851.1 | - | 15 - 16 | 1935 | 105 | ||
AK155900.1 | - | 15 - 16 | 1935 | 105 | ||
AK171319.1 | - | 15 - 16 | 1936 | 105 | ||
BC119222.1 | - | 15 - 16 | 1869 | 8733971 | 105 | |
BC120545.1 | - | 15 - 16 | 1869 | 40129468 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |