Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190978.1 | NP_001177907.1 | 2 - 3 | 632 | 60 | |
NM_010623.4 | NP_034753.1 | 2 - 3 | 632 | 60 | ||
GenBank mRNA | AB001424.1 | - | 1 - 2 | 122 | 60 | |
AB008867.1 | - | 2 - 3 | 630 | 60 | ||
AK046890.1 | - | 2 - 3 | 385 | 60 | ||
AK077868.1 | - | 2 - 3 | 413 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |