Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011270.3 | NP_035400.3 | 4 - 5 | 714 | 59 | |
GenBank mRNA | AB015189.1 | - | 4 - 5 | 649 | 59 | |
AF012430.1 | - | 4 - 5 | 629 | 59 | ||
AF047827.1 | - | 4 - 5 | 649 | 59 | ||
AF057524.1 | - | 4 - 5 | 666 | 59 | ||
AF069311.1 | - | 4 - 5 | 668 | 59 | ||
AK089642.1 | - | 4 - 5 | 667 | 59 | ||
AK157850.1 | - | 4 - 5 | 641 | 59 | ||
AK166030.1 | - | 2 - 3 | 397 | 59 | ||
AK171641.1 | - | 4 - 5 | 655 | 59 | ||
BC145805.1 | - | 4 - 5 | 745 | 40131108 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |