Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243047.1 | NP_001229976.1 | 5 - 6 | 670 | 79 | |
NM_011807.3 | NP_035937.2 | 17 - 18 | 2146 | 79 | ||
GenBank mRNA | AF388675.1 | - | 17 - 18 | 1925 | 79 | |
AK039754.1 | - | 5 - 6 | 535 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |