Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013807.2 | NP_038835.2 | 1 - 2 | 300 | 73 | |
GenBank mRNA | BC031180.1 | - | 1 - 2 | 292 | 4988360 | 73 |
BC063051.1 | - | 1 - 2 | 300 | 5718130 | 73 | |
U21392.1 | - | 1 - 2 | 147 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |