Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016979.1 | NP_058675.1 | 3 - 4 | 840 | 148 | |
GenBank mRNA | AJ238004.1 | - | 3 - 4 | 723 | 148 | |
AK034681.1 | - | 3 - 4 | 1159 | 148 | ||
AK036432.1 | - | 3 - 4 | 805 | 148 | ||
AK037141.1 | - | 3 - 4 | 834 | 148 | ||
AK039088.1 | - | 3 - 4 | 855 | 148 | ||
AK081548.1 | - | 3 - 4 | 820 | 148 | ||
AK139510.1 | - | 3 - 4 | 848 | 148 | ||
AK150588.1 | - | 3 - 4 | 802 | 148 | ||
AK152166.1 | - | 3 - 4 | 637 | 148 | ||
AK154447.1 | - | 3 - 4 | 820 | 148 | ||
AK169322.1 | - | 3 - 4 | 791 | 148 | ||
BC006875.1 | - | 3 - 4 | 840 | 3596013 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus 1700 Probe spans exons |