Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900015003.mm.1 |
NM_018831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015003.mm.1 |
NM_018831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015003.mm.1 |
TC1900001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015005.mm.1 |
NM_018831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015005.mm.1 |
NM_018831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015005.mm.1 |
TC1900001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015002.mm.1 |
NM_018831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015002.mm.1 |
NM_018831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015002.mm.1 |
TC1900001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015004.mm.1 |
NM_018831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015004.mm.1 |
NM_018831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015004.mm.1 |
TC1900001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015006.mm.1 |
NM_018831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015006.mm.1 |
NM_018831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015006.mm.1 |
TC1900001705.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900015010.mm.1 |
NM_018831.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015010.mm.1 |
NM_018831.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900015010.mm.1 |
TC1900001705.mm.1 |
MTA TC |