Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019546.5 | NP_062419.2 | 7 - 8 | 1020 | 67 | |
GenBank mRNA | AF222851.1 | - | 7 - 8 | 1007 | 67 | |
AK011020.1 | - | 7 - 8 | 1012 | 67 | ||
AK050141.1 | - | 7 - 8 | 1021 | 67 | ||
BC018182.1 | - | 7 - 8 | 1006 | 4161894 | 67 | |
U80019.1 | - | 7 - 8 | 1018 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |