Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199275.2 | NP_001186204.1 | 7 - 8 | 966 | 111 | |
NM_001199276.2 | NP_001186205.1 | 5 - 6 | 636 | 111 | ||
NM_001271455.1 | NP_001258384.1 | 6 - 7 | 689 | 111 | ||
NM_001271456.1 | NP_001258385.1 | 7 - 8 | 861 | 111 | ||
NM_021327.4 | NP_067302.2 | 6 - 7 | 808 | 111 | ||
GenBank mRNA | AJ242777.1 | - | 5 - 6 | 608 | 111 | |
AJ242778.1 | - | 6 - 7 | 780 | 111 | ||
AK135338.1 | - | 6 - 7 | 787 | 111 | ||
AK156131.1 | - | 6 - 7 | 661 | 111 | ||
AK170841.1 | - | 5 - 6 | 614 | 111 | ||
AK172890.1 | - | 594 | 111 | |||
AY012159.2 | - | 3 - 4 | 378 | 111 | ||
BC008186.1 | - | 6 - 7 | 725 | 3589311 | 111 | |
BC008665.1 | - | 7 - 8 | 832 | 3598934 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |