Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700004540.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004540.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004532.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004532.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004533.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004533.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004537.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004537.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004542.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004542.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004535.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004535.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004524.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004524.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004529.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004529.mm.1 |
TC1700002773.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700004531.mm.1 |
NM_021419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700004531.mm.1 |
TC1700002773.mm.1 |
MTA TC |