Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53inp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199105.1 | NP_001186034.1 | 1 - 2 | 110 | 106 | |
NM_021897.3 | NP_068697.1 | 1 - 2 | 110 | 106 | ||
GenBank mRNA | AJ131776.1 | - | 1 - 2 | 111 | 106 | |
AK054214.1 | - | 1 - 2 | 108 | 106 | ||
AK142321.1 | - | 1 - 2 | 106 | 106 | ||
AY034611.1 | - | 1 - 2 | 111 | 106 | ||
AY034612.1 | - | 1 - 2 | 111 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons |