Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164791.1 | NP_001158263.1 | 10 - 11 | 1035 | 74 | |
NM_023057.5 | NP_075544.1 | 10 - 11 | 1099 | 74 | ||
GenBank mRNA | AB049731.1 | - | 9 - 10 | 882 | 74 | |
AB049732.1 | - | 9 - 10 | 882 | 74 | ||
AK087363.1 | - | 10 - 11 | 975 | 74 | ||
AK132186.1 | - | 10 - 11 | 1100 | 74 | ||
AK162819.1 | - | 10 - 11 | 900 | 74 | ||
BC023718.1 | - | 10 - 11 | 980 | 5346593 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |