Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023626.4 | NP_076115.3 | 9 - 10 | 1058 | 89 | |
GenBank mRNA | AK030364.1 | - | 8 - 9 | 865 | 89 | |
AK080787.1 | - | 9 - 10 | 1027 | 89 | ||
AK131993.1 | - | 9 - 10 | 1058 | 89 | ||
AK146145.1 | - | 8 - 9 | 841 | 89 | ||
AY007791.1 | - | 9 - 10 | 1027 | 89 | ||
BC005721.1 | - | 2 - 3 | 247 | 89 | ||
BC018342.1 | - | 9 - 10 | 1008 | 4010253 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |