Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ush1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF228924.1 | - | 10 - 11 | 850 | 67 | |
AF228925.1 | - | 10 - 11 | 850 | 67 | ||
AY103465.1 | - | 10 - 11 | 850 | 67 | ||
AY246556.1 | - | 10 - 11 | 824 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |