Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025948.2 | NP_080224.1 | 2 - 3 | 348 | 83 | |
GenBank mRNA | AK012276.1 | - | 1 - 2 | 111 | 83 | |
AK189673.1 | - | 1 - 2 | 83 | 83 | ||
BC031521.1 | - | 2 - 3 | 348 | 2655161 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |