Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sncaip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199151.1 | NP_001186080.1 | 7 - 8 | 1599 | 66 | |
NM_001199153.1 | NP_001186082.1 | 7 - 8 | 1504 | 66 | ||
NM_001199154.1 | NP_001186083.1 | 7 - 8 | 1774 | 66 | ||
NM_026408.4 | NP_080684.3 | 7 - 8 | 1599 | 66 | ||
GenBank mRNA | AF348447.1 | - | 7 - 8 | 1527 | 66 | |
AK013040.1 | - | 5 - 6 | 1333 | 66 | ||
AK017012.1 | - | 4 - 5 | 534 | 66 | ||
AK044974.1 | - | 7 - 8 | 1797 | 66 | ||
AK081942.1 | - | 7 - 8 | 1778 | 66 | ||
AK082079.1 | - | 7 - 8 | 1603 | 66 | ||
AK132346.1 | - | 7 - 8 | 1702 | 66 | ||
AK160111.1 | - | 7 - 8 | 1603 | 66 | ||
BC098489.1 | - | 7 - 8 | 1503 | 30096466 | 66 | |
BC130225.1 | - | 7 - 8 | 1489 | 66 | ||
BC152544.1 | - | 7 - 8 | 1487 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |