Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Steap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027399.3 | NP_081675.2 | 1 - 2 | 71 | 79 | |
GenBank mRNA | AK010437.1 | - | 1 - 2 | 66 | 79 | |
AK144312.1 | - | 1 - 2 | 26 | 79 | ||
BC061023.1 | - | 1 - 2 | 56 | 30304473 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |