Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100015886.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015886.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015877.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015877.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015869.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015869.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015873.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015873.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015881.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015881.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015883.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015883.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015870.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015870.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015874.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015874.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015885.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015885.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015887.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015887.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015876.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015876.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015880.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015880.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015884.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015884.mm.1 |
TC0100001932.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100015888.mm.1 |
NM_027671.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100015888.mm.1 |
TC0100001932.mm.1 |
MTA TC |