Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200031010.mm.1 |
NM_001164477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031010.mm.1 |
NM_001164477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031010.mm.1 |
NM_027835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031010.mm.1 |
NM_027835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031010.mm.1 |
TC0200003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031011.mm.1 |
NM_001164477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031011.mm.1 |
NM_001164477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031011.mm.1 |
NM_027835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031011.mm.1 |
NM_027835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031011.mm.1 |
TC0200003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031014.mm.1 |
NM_001164477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031014.mm.1 |
NM_001164477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031014.mm.1 |
NM_027835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031014.mm.1 |
NM_027835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031014.mm.1 |
TC0200003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031015.mm.1 |
NM_001164477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031015.mm.1 |
NM_001164477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031015.mm.1 |
NM_027835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031015.mm.1 |
NM_027835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031015.mm.1 |
TC0200003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031009.mm.1 |
NM_027835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031009.mm.1 |
NM_027835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031009.mm.1 |
TC0200003630.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200031012.mm.1 |
NM_001164477.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031012.mm.1 |
NM_001164477.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031012.mm.1 |
NM_027835.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031012.mm.1 |
NM_027835.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200031012.mm.1 |
TC0200003630.mm.1 |
MTA TC |