Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029746.3 | NP_084022.2 | 8 - 9 | 1007 | 84 | |
GenBank mRNA | AK089920.1 | - | 8 - 9 | 991 | 84 | |
AK157348.1 | - | 8 - 9 | 969 | 84 | ||
AK169298.1 | - | 8 - 9 | 977 | 84 | ||
AK171726.1 | - | 8 - 9 | 926 | 84 | ||
BC011435.1 | - | 8 - 9 | 947 | 4010102 | 84 | |
CO423795.1 | - | 5 - 6 | 564 | 84 | ||
CX205121.1 | - | 2 - 3 | 216 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |