Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029746.3 | NP_084022.2 | 11 - 12 | 1347 | 72 | |
GenBank mRNA | AK089920.1 | - | 11 - 12 | 1331 | 72 | |
AK157348.1 | - | 11 - 12 | 1309 | 72 | ||
AK169298.1 | - | 11 - 12 | 1317 | 72 | ||
AK171726.1 | - | 11 - 12 | 1266 | 72 | ||
AK189695.1 | - | 2 - 3 | 146 | 72 | ||
BC011435.1 | - | 11 - 12 | 1287 | 4010102 | 72 | |
CX205121.1 | - | 5 - 6 | 556 | 72 | ||
W74920.1 | - | 2 - 3 | 232 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |