Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164634.1 | NP_001158106.1 | 8 - 9 | 1443 | 89 | |
NM_001164635.1 | NP_001158107.1 | 8 - 9 | 1351 | 89 | ||
NM_031194.5 | NP_112471.3 | 9 - 10 | 1523 | 89 | ||
GenBank mRNA | AB079895.1 | - | 7 - 8 | 1180 | 89 | |
AF078869.1 | - | 8 - 9 | 1298 | 89 | ||
AK031962.1 | - | 9 - 10 | 1489 | 89 | ||
AK044336.1 | - | 8 - 9 | 1320 | 89 | ||
BC014762.1 | - | 8 - 9 | 1329 | 4239544 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |