Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033759.2 | NP_001028931.1 | 10 - 11 | 2508 | 85 | |
NM_031997.4 | NP_114386.3 | 11 - 12 | 2590 | 85 | ||
GenBank mRNA | AK129352.1 | - | 5 - 6 | 809 | 85 | |
BC019745.1 | - | 2 - 3 | 231 | 3675225 | 85 | |
BC076570.1 | - | 10 - 11 | 2114 | 6830791 | 85 | |
BC089353.1 | - | 10 - 11 | 2490 | 6410437 | 85 | |
CA752446.1 | - | 3 - 4 | 448 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |