Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145800.1 | NP_001139272.1 | 13 - 14 | 1899 | 92 | |
NM_033608.3 | NP_291086.2 | 13 - 14 | 1965 | 92 | ||
GenBank mRNA | AF317839.1 | - | 13 - 14 | 1965 | 92 | |
AK078536.1 | - | 13 - 14 | 1893 | 92 | ||
AK145121.1 | - | 13 - 14 | 1874 | 92 | ||
BC040281.1 | - | 13 - 14 | 1925 | 5364073 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |