Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmynd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053253.3 | NP_444483.2 | 5 - 6 | 655 | 64 | |
GenBank mRNA | AF123386.1 | - | 3 - 4 | 64 | ||
AK132332.1 | - | 5 - 6 | 664 | 64 | ||
BC052357.1 | - | 5 - 6 | 655 | 6772144 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |