Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Def8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253783.1 | NP_001240712.1 | 10 - 11 | 1126 | 76 | |
NM_001253784.1 | NP_001240713.1 | 9 - 10 | 1104 | 76 | ||
NM_001281803.1 | NP_001268732.1 | 8 - 9 | 1108 | 76 | ||
NM_054046.5 | NP_473387.1 | 8 - 9 | 1017 | 76 | ||
GenBank mRNA | AK028726.1 | - | 9 - 10 | 1076 | 76 | |
AK034028.1 | - | 983 | 76 | |||
AK046271.1 | - | 8 - 9 | 977 | 76 | ||
AK050405.1 | - | 9 - 10 | 1075 | 76 | ||
AK081295.1 | - | 8 - 9 | 1007 | 76 | ||
AK086174.1 | - | 8 - 9 | 980 | 76 | ||
AK087778.1 | - | 8 - 9 | 988 | 76 | ||
AK133979.1 | - | 8 - 9 | 991 | 76 | ||
AK150334.1 | - | 8 - 9 | 953 | 76 | ||
AK150994.1 | - | 8 - 9 | 953 | 76 | ||
AK164147.1 | - | 8 - 9 | 989 | 76 | ||
BC003306.1 | - | 10 - 11 | 1077 | 3154712 | 76 | |
CD545768.1 | - | 4 - 5 | 503 | 76 | ||
CN705834.1 | - | 1 - 2 | 58 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |