Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF169620.1 | - | 4 - 5 | 440 | 69 | |
AK077456.1 | - | 4 - 5 | 497 | 69 | ||
AK140200.1 | - | 4 - 5 | 524 | 69 | ||
AK144412.1 | - | 4 - 5 | 495 | 69 | ||
AK145829.1 | - | 4 - 5 | 497 | 69 | ||
AK146754.1 | - | 4 - 5 | 496 | 69 | ||
AK146966.1 | - | 4 - 5 | 504 | 69 | ||
AK159186.1 | - | 4 - 5 | 503 | 69 | ||
BC088997.1 | - | 4 - 5 | 454 | 69 | ||
BC122563.1 | - | 3 - 4 | 428 | 40107557 | 69 | |
BC125275.1 | - | 3 - 4 | 428 | 40107558 | 69 | |
BY727303.1 | - | 4 - 5 | 523 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |