Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpnpep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289729.1 | NP_001276658.1 | 19 - 20 | 2359 | 71 | |
NM_133213.2 | NP_573476.2 | 19 - 20 | 2158 | 71 | ||
GenBank mRNA | AF367247.1 | - | 19 - 20 | 1822 | 71 | |
AF428102.1 | - | 19 - 20 | 2009 | 71 | ||
AK078835.1 | - | 19 - 20 | 1965 | 71 | ||
AK085520.1 | - | 19 - 20 | 2157 | 71 | ||
AK164538.1 | - | 19 - 20 | 1986 | 71 | ||
AK172086.1 | - | 19 - 20 | 2159 | 71 | ||
BC140977.1 | - | 19 - 20 | 1802 | 9055589 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |