Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134154.3 | NP_598915.1 | 3 - 4 | 341 | 101 | |
GenBank mRNA | AK133879.1 | - | 3 - 4 | 995 | 101 | |
AK155754.1 | - | 3 - 4 | 994 | 101 | ||
AK169876.1 | - | 3 - 4 | 987 | 101 | ||
AK170869.1 | - | 4 - 5 | 472 | 101 | ||
BC022156.1 | - | 3 - 4 | 172 | 101 | ||
BC037680.1 | - | 3 - 4 | 345 | 3481846 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |