Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcoln3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134160.1 | NP_598921.1 | 4 - 5 | 639 | 90 | |
GenBank mRNA | AF475086.1 | - | 4 - 5 | 639 | 90 | |
AK030819.1 | - | 4 - 5 | 759 | 90 | ||
AK033008.1 | - | 4 - 5 | 753 | 90 | ||
AK035029.1 | - | 4 - 5 | 760 | 90 | ||
AY083531.1 | - | 4 - 5 | 637 | 90 | ||
BC106856.2 | - | 4 - 5 | 639 | 40047039 | 90 | |
BC106857.2 | - | 4 - 5 | 639 | 40047042 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |