Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200014966.mm.1 |
NM_139308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014966.mm.1 |
NM_139308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014966.mm.1 |
TC0200001911.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014962.mm.1 |
NM_139308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014962.mm.1 |
NM_139308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014962.mm.1 |
TC0200001911.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014963.mm.1 |
NM_139308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014963.mm.1 |
NM_139308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014963.mm.1 |
TC0200001911.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014964.mm.1 |
NM_139308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014964.mm.1 |
NM_139308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014964.mm.1 |
TC0200001911.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014961.mm.1 |
NM_139308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014961.mm.1 |
NM_139308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014961.mm.1 |
TC0200001911.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200014965.mm.1 |
NM_139308.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014965.mm.1 |
NM_139308.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200014965.mm.1 |
TC0200001911.mm.1 |
MTA TC |