Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168256.1 | NP_001161728.1 | 6 - 7 | 572 | 97 | |
NM_001168257.1 | NP_001161729.1 | 5 - 6 | 497 | 97 | ||
NM_144805.2 | NP_659054.2 | 6 - 7 | 440 | 97 | ||
GenBank mRNA | BC019416.1 | - | 6 - 7 | 402 | 5151159 | 97 |
CT010303.1 | - | 5 - 6 | 370 | 97 | ||
CV561368.1 | - | 5 - 6 | 439 | 97 | ||
CX564997.1 | - | 6 - 7 | 471 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |