Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144856.2 | NP_659105.2 | 2 - 3 | 590 | 63 | |
GenBank mRNA | AK082865.1 | - | 2 - 3 | 591 | 63 | |
AK143949.1 | - | 2 - 3 | 578 | 63 | ||
BC013474.1 | - | 2 - 3 | 567 | 4236556 | 63 | |
BC024119.1 | - | 2 - 3 | 554 | 5096875 | 63 | |
BC025813.1 | - | 2 - 3 | 221 | 5097639 | 63 | |
BC026597.1 | - | 2 - 3 | 544 | 4975360 | 63 | |
BC026598.1 | - | 2 - 3 | 542 | 4975598 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |