Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lppr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290299.1 | NP_001277228.1 | 4 - 5 | 586 | 84 | |
NM_144935.1 | NP_659184.1 | 3 - 4 | 558 | 84 | ||
GenBank mRNA | AK082453.1 | - | 4 - 5 | 549 | 84 | |
AY438023.1 | - | 3 - 4 | 558 | 84 | ||
BC018242.1 | - | 3 - 4 | 558 | 4503683 | 84 | |
BC023082.1 | - | 4 - 5 | 563 | 5357160 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |