Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neto1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144946.4 | NP_659195.3 | 6 - 7 | 1566 | 77 | |
GenBank mRNA | AF448840.1 | - | 6 - 7 | 899 | 77 | |
AK032510.1 | - | 4 - 5 | 434 | 77 | ||
AK043710.1 | - | 6 - 7 | 902 | 77 | ||
AK081325.1 | - | 6 - 7 | 1567 | 77 | ||
AY138990.1 | - | 6 - 7 | 635 | 77 | ||
BC051145.1 | - | 6 - 7 | 926 | 6590345 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |