Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145226.2 | NP_660261.1 | 4 - 5 | 907 | 60 | |
GenBank mRNA | AB067534.1 | - | 4 - 5 | 907 | 60 | |
AF453830.1 | - | 4 - 5 | 899 | 60 | ||
AK158392.1 | - | 4 - 5 | 889 | 60 | ||
BC141055.1 | - | 4 - 5 | 900 | 9055667 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |