Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145488.1 | NP_663463.1 | 9 - 10 | 2008 | 118 | |
GenBank mRNA | AK143173.1 | - | 9 - 10 | 1996 | 118 | |
AK220525.1 | - | 9 - 10 | 1989 | 118 | ||
BC003424.1 | - | 9 - 10 | 2008 | 2609884 | 118 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |