Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lctl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145835.2 | NP_665834.1 | 9 - 10 | 1193 | 83 | |
GenBank mRNA | AF309072.1 | - | 9 - 10 | 1194 | 83 | |
BC030631.1 | - | 5 - 6 | 518 | 1195880 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |